Orders LinkContactUsLinkComponent
Microscope objective lens mounts
Microscope objective lens mounts
Supplier:   DIDALAB
Microscope objective lens mounts
Supplier:  DIDALAB
Certificates
Microscope objective lens mounts
Microscope objective lens mounts
2 Options Available Below
Product Details & Documents
The objective holder allows precise positioning of a microscope objective according to the vertical and the horizontal.
  • Both axes 2 mm each
  • Holder is mounted on a 10 mm diameter rod
Product Family Options
Product Information
  • Availability
    Price
  • DIDAPOD060550
    Lens holder with X/Y adjustment
    Lens holder with X/Y adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,88 €
    91,88 €
    1 * 1 ST
       91,88 €
  • DIDAPOD060560
    Lens holder with angular adjustment
    Lens holder with angular adjustment
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,88 €
    91,88 €
    1 * 1 ST
       91,88 €