Orders LinkContactUsLinkComponent
Capillary columns, TracePLOT™ TG-BOND U
Capillary columns, TracePLOT™ TG-BOND U
Capillary columns, TracePLOT™ TG-BOND U
Certificates
Capillary columns, TracePLOT™ TG-BOND U
3 Options Available Below
Product Details & Documents
TracePLOT™ TG-BOND U GC columns are ideal for the analysis of polar compounds.
  • Polar, divinylbenzene ethylene glycol/dimethylacrylate phase
  • Particles incorporated into the walls of the tubing for essentially no particle release
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TG-BOND U
    10,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 778,00 €
    778,00 €
    1 * 1 ST
       778,00 €
  • Capillary
    TG-BOND U
    10,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • Capillary
    TG-BOND U
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 170,00 €
    1 170,00 €
    1 * 1 ST
       1 170,00 €