Sealing film, self-adhesive for automation
391-0700
: Brand
- Description:Sealing film, self-adhesive, for automation, PP/PE, single sheets
- Pk:50
Chemical resistant PE/PP film offers adhesive-free areas over the wells to avoid contamination. Furthermore, it can be pierced easily with a pipette tip (for sample removal). Temperature range: −40 to +90 °C.