Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
Catalog # 732-1475
Supplier: Brand
Specifications
- Optical property:Transparent
- Description:Sealing film, self-adhesive, for ELISA, Deep Well and PCR plates, polyester, single sheets
- Pk:100
Specifications
About this item
Highly transparent film made of polyester is suitable for Real-Time PCR measurements. Temperature range: −40 to +120 °C.