Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification and general filtration.
- Supor® membrane offers optimal support to retain chromatography resins whilst allowing smooth flow of buffers; Mustang® membrane is able to withstand high flow rates to render fast purification of biomolecules
- Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume
- Outlet tip geometry minimises sample leakage and loss during incubation steps
- Low non specific binding - intrinsic plate properties prevent target molecules from binding to the plate
Applications for AcroPrep™ Advance 96-well filter plates for protein purification include high throughput protein purification, screening of chromatography resins, screening of chromatography conditions, protein fractionation, and antibody purification.
Related Information
Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Application Notes
Pall Laboratory AcroPrep Filter-Plates
Product Literature
Frequently Bought Together