Order Entry
Belgium
ContactUsLinkComponent
Forestry safety helmet, combi kit, PW98
Forestry safety helmet, combi kit, PW98
Catalog # PRTWPW98ORR
Supplier:  Portwest
undefined
Forestry safety helmet, combi kit, PW98
Catalog # PRTWPW98ORR
Supplier:  Portwest

Specifications

  • Colour:
    Orange
  • Description:
    Forestry head protection kit, with visor, hearing protection, sun peak, neck guard
  • Pk:
    1

Specifications

About this item

This professional head protection kit comes compete with visor hearing protection, sun peak and neck guard (ABS helmet, mesh visor, ABS/polystyrene ear muffs).

  • Class1 optics and high tech design provide good visibility and full face protection
  • Kit is completed with a reinforced visor holder, increasing the stability of the helmet
  • Ear protection: SNR23dB

Colour: Orange

Certifications: EN 397, EN 352-3, EN 166 3B, EN 1731