Orders LinkContactUsLinkComponent
Wheaton® MicroLiter™ plate sampling system
Wheaton® MicroLiter™ plate sampling system
Wheaton® MicroLiter™ plate sampling system
Certificates
Wheaton® MicroLiter™ plate sampling system
Wheaton® MicroLiter™ plate sampling system
6 Options Available Below
Product Details & Documents
A comprehensive range of microplates, glass inserts, sealing mats and/or caps with chromatography-based septa that offer security for even the most volatile samples.
Polypropylene μLplates™ have a variety of closures that will stabilise the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. There is the option of choosing the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system permits even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/silicone rubber using a mechanical seal. A variety of insert options are available for the appropriate application.
96-well round and square well microplates are available in a variety of volumes and options. Made of natural PP, they are made to ANSI specifications for 96-well microplates.
96-well round and square well covers are made of Ethylene Vinyl Acetate (EVA) and are pierceable by standard needle configurations found on most HPLC, LC/MS, CombiChem, and HTS instruments. They have a superior seal compared to similar products. For users who require some level of resealability, two silicone mats with a sprayed-on fluoropolymer resin barrier are available. Silicone mats are available in standard and pre-cut round or square well formats.
Product Family Options
Plates, μLplate®
Product Information
  • Well volume
    Availability
    Price
  • MLAS07-8000
    μLplate®, 96 deep well, PP, square well, round bottom
    μLplate®, 96 deep well, PP, square well, round bottom
    2000 µl
    Discontinued
  • MLAS07-8400
    μLplate®, 96 deep well, PP, natural, square well, round bottom
    μLplate®, 96 deep well, PP, natural, square well, round bottom
    2000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Glass inserts
Product Information
  • Well volume
    Availability
    Price
  • MLAS07-0000-2200
    Glass inserts, 2,2 ml, clear glass, round, for 96 deep well μLplate® (2000 µl)
    Glass inserts, 2,2 ml, clear glass, round, for 96 deep well μLplate® (2000 µl)
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
µLplate® with glass inserts installed
Product Information
  • Well volume
    Availability
    Price
  • MLAS07-8000-1200
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted)
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted)
    2000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • MLAS07-8000-2200
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted)
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted)
    2000 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
µLplate® with glass inserts installed and a detached μLmat™
Product Information
  • Well volume
    Availability
    Price
  • MLAS078020MB1200
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • MLAS078020MB2200
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Snap caps and μLMATS™
Product Information
  • Catalog #
    Description
    Availability
    Price
  • μLmat™, blue, with pre-cut FEP/silicone septa
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST
       0,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Snap cap, blue, with pre-cut FEP/silicone septa
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 0,00 €
    0,00 €
    1 * 100 ST
       0,00 €
Covers and silicone mats
Product Information
  • Catalog #
    Description
    Availability
    Price
  • EVA plate cover, natural, pierceable, for square well μLplates®
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST