Safety ankle boots, lace-up, Steelite™ WORK Thor FW11
Supplier: Portwest
Certificates
About this item
Anti-static safety ankle boots made from full grain water resistant leather and a dual-density PU/TPU sole (petrol and chemical resistant).
- Shock absorbent heel
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
Colour: Black
Certifications: EN ISO 20345:2011; S3
Frequently Bought Together