Orders LinkContactUsLinkComponent
Handheld refractometer, OE 200
Handheld refractometer, OE 200
Handheld refractometer, OE 200, 0 - 200 Oe° ; 0 - 45 °Bx ; 0 - 40 KI°, 1,3330 - 1,4100 nD
Catalog # 635-2115
Supplier:   REICHERT
Handheld refractometer, OE 200
Catalog # 635-2115
Supplier:  REICHERT
Handheld refractometer, OE 200
Handheld refractometer, OE 200
Handheld refractometer, OE 200, 0 - 200 Oe° ; 0 - 45 °Bx ; 0 - 40 KI°, 1,3330 - 1,4100 nD
Price
338,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This handheld refractometer for wine provides fast and accurate measurements of dissolved sugars when testing grapes, citrus fruits, fruit juices, soft drinks, melons and maple sap for the winemaking industry.
Specifications
  • Model:
    OE 200
  • Range:
    0 - 200 Oe° ; 0 - 45 °Bx ; 0 - 40 KI°
  • Description:
    Handheld refractometer
  • Refractive index:
    1,3330 - 1,4100 nD
  • Cat. No.:
    635-2115
  • Scales:
    Oechsle (Oeo), Brix (% sucrose) and Klosterneuburger (Klo)
  • Temperature compensation range:
    10...40 °C