Orders LinkContactUsLinkComponent
m-ColiBlue24® broth
m-ColiBlue24® broth
Supplier:   Hach
m-ColiBlue24® broth
Supplier:  Hach
Certificates
m-ColiBlue24® broth
m-ColiBlue24® broth
3 Options Available Below
Product Details & Documents
In 24 hours, Hach's m-ColiBlue24® broth give the simultaneous results for total coliforms and E. coli. Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media. Glass ampules, plastic ampules, 100 ml bottles of m-ColiBlue24® and agar plates, afford maximum shelf life and ease of use for pre-measured and prepared media. Enumerate total coliforms and E. coli on one petri dish. Differentiate colonies easily - red and blue indicate total coliforms and blue specifies E. coli. Get optimal recovery of stressed and injured organisms. Use m-ColiBlue24® broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters. With ready-to-use m-ColiBlue24® Broth, you can:
  • Simultaneously detect total coliforms and E. coli in just 24 hours
  • Easily read and interpret results
  • Save money by eliminating special equipment and UV lamps
  • Minimize background growth of non-coliform bacteria, reduce false positives and false negatives, plus get superior sensitivity down to 1 CFU/100 ml
  • Conduct emergency testing and routine monitoring
m-ColiBlue24® should only be used with pads manufactured by the following companies/brands: Pall Corporation (e.g., sterile petri dishes with absorbent pads), Private Label/OEM products manufactured by Pall Gelman, Sartorius (cellulose and glass fibre pads), and Sartorius BioSart products with the integrated Petri dish. No other pad is compatible with the product, and the use of any pad other than those recommended may result in unreliable, inaccurate data.
Documents
Product Family Options
Product Information
  • For
    Pk
    Availability
    Price
  • HACH2608420
    m-ColiBlue24® Broth, glass ampules
    m-ColiBlue24® Broth, glass ampules
    E.coli
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 86,90 €
    86,90 €
    1 * 20 ST
       86,90 €
  • HACH2608450
    m-ColiBlue24® Broth, plastic ampules
    m-ColiBlue24® Broth, plastic ampules
    E.coli
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 191,00 €
    191,00 €
    1 * 50 ST
       191,00 €
  • HACH2608442
    m-ColiBlue24® Broth, bottle, 100 ml
    m-ColiBlue24® Broth, bottle, 100 ml
    E.coli
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 106,00 €
    106,00 €
    1 * 100 mL
       106,00 €