Orders LinkContactUsLinkComponent
Test tubes with screw caps
Test tubes with screw caps
Test tubes with screw caps
Supplier:  BOETTGER PAUL
Certificates
Test tubes with screw caps
Test tubes with screw caps
10 Options Available Below
Product Details & Documents
PP, transparent.
  • Good chemical and temperature resistance
  • Tight sealing screw cap made from PE
  • Available with enclosed or fitted screw cap, standard-hygienic or sterile
For safe transportation and storage of samples.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Cap colour
    Type
    Availability
    Price
  • 212-0517
    Flat bottom, screw cap enclosed
    Flat bottom, screw cap enclosed
    8 ml
    57 mm
    16 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 297,00 €
    297,00 €
    1 * 2.000 ST
       297,00 €
  • 212-0511
    Conical bottom, skirted, screw cap enclosed
    Conical bottom, skirted, screw cap enclosed
    15 ml
    76 mm
    20 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 180,00 €
    180,00 €
    1 * 1.000 ST
       180,00 €
  • 212-0512
    Conical bottom, skirted, screw cap fitted
    Conical bottom, skirted, screw cap fitted
    15 ml
    76 mm
    20 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 231,00 €
    231,00 €
    1 * 1.000 ST
       231,00 €
  • 212-0513
    Conical bottom, skirted, screw cap fitted
    Conical bottom, skirted, screw cap fitted
    15 ml
    76 mm
    20 mm
    Natural
    sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 320,00 €
    320,00 €
    1 * 1.000 ST
       320,00 €
  • 212-0508
    Round bottom, screw cap enclosed
    Round bottom, screw cap enclosed
    13 ml
    100 mm
    16 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 145,00 €
    145,00 €
    1 * 1.000 ST
       145,00 €
  • 212-0509
    Round bottom, screw cap fitted
    Round bottom, screw cap fitted
    13 ml
    100 mm
    16 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 202,00 €
    202,00 €
    1 * 1.000 ST
       202,00 €
  • 212-0510
    Round bottom, screw cap fitted
    Round bottom, screw cap fitted
    13 ml
    100 mm
    16 mm
    Natural
    sterile
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 289,00 €
    289,00 €
    1 * 1.000 ST
       289,00 €
  • 212-0514
    Conical bottom, skirted, screw cap enclosed
    Conical bottom, skirted, screw cap enclosed
    30 ml
    107 mm
    25 mm
    Natural
    non sterile, graduated
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 229,00 €
    229,00 €
    1 * 500 ST
       229,00 €
  • 212-0515
    Conical bottom, skirted, screw cap fitted
    Conical bottom, skirted, screw cap fitted
    30 ml
    107 mm
    25 mm
    Natural
    non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 259,00 €
    259,00 €
    1 * 500 ST
       259,00 €
  • 212-0516
    Conical bottom, skirted, screw cap fitted
    Conical bottom, skirted, screw cap fitted
    30 ml
    107 mm
    25 mm
    Natural
    sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 318,00 €
    318,00 €
    1 * 500 ST
       318,00 €