Luminescent/optical dissolved oxygen (LDO) electrodes, IntelliCAL™
Supplier: Hach
Jump to:
About this item
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
- Rugged digital probes can reliably test in the harshest environmental conditions: Stainless steel body, polymer sensor shroud, and reinforced, steel-sheathed cables protect the electrode for measurement anywhere in the field
- No membranes to clean or replace, no electrolyte solution to replenish, and reduce calibration frequency
- Ultimate traceability: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
- Alert the user when re-calibration is needed
- Electrodes can be moved between meters without the need to re-calibrate or re-enter measurement settings
- Ideal for a multi-user environment with multiple HQd series meters
The standard version is intended for laboratory use, the rugged version is intended for field use.
6 Options Available Below
Product Details & Documents
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
- Rugged digital probes can reliably test in the harshest environmental conditions: Stainless steel body, polymer sensor shroud, and reinforced, steel-sheathed cables protect the electrode for measurement anywhere in the field
- No membranes to clean or replace, no electrolyte solution to replenish, and reduce calibration frequency
- Ultimate traceability: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
- Alert the user when re-calibration is needed
- Electrodes can be moved between meters without the need to re-calibrate or re-enter measurement settings
- Ideal for a multi-user environment with multiple HQd series meters
The standard version is intended for laboratory use, the rugged version is intended for field use.
Documents
Product Family Options
Standard LDO electrodes, IntelliCAL™
Product Information
- TypeConcentrationTemp.MaterialConnectionØ×LForAvailabilityPrice
- 664-0206IntelliCAL™ LDO101, standard LDO electrodeIntelliCAL™ LDO101, standard LDO electrodeIntelliCAL™ LDO101, standard0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABSM12, 1 m cable15×200 mmWastewater, drinking water and general water quality applicationsLog in for availability1 * 1 ST1 140,00 €
- 664-0207IntelliCAL™ LDO101, standard LDO electrodeIntelliCAL™ LDO101, standard LDO electrodeIntelliCAL™ LDO101, standard0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABSM12, 3 m cable15×200 mmWastewater, drinking water and general water quality applicationsLog in for availability1 * 1 ST1 140,00 €
Rugged LDO electrodes for outdoor use, IntelliCAL™
Product Information
- TypeConcentrationTemp.MaterialConnectionØ×LForAvailabilityPrice
- 664-0208IntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABS/stainless steelM12, 5 m cable45×250 mmWastewater, drinking water and general water quality applications (field use)Log in for availability1 * 1 ST1 360,00 €
- 664-0209IntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABS/stainless steelM12, 10 m cable45×250 mmWastewater, drinking water and general water quality applications (field use)Log in for availability1 * 1 ST1 380,00 €
- 664-0210IntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABS/stainless steelM12, 15 m cable45×250 mmWastewater, drinking water and general water quality applications (field use)Log in for availability1 * 1 ST1 390,00 €
- 664-0211IntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged LDO electrodeIntelliCAL™ LDO101, rugged0,1 - 20 mg/l (1 - 200%)0...+50 °CPC/ABS/stainless steelM12, 30 m cable45×250 mmWastewater, drinking water and general water quality applications (field use)Log in for availability1 * 1 ST1 430,00 €
Accessories
Product Information
- Catalog #CapacityAvailabilityPrice
- BOD bottle
Product Information
- Catalog #AvailabilityPrice
- Protective shroud kit for standard IntelliCAL™ LDO probes (incl. shroud and locking collar)Protective shroud kit for standard IntelliCAL™ LDO probes (incl. shroud and locking collar)HACH5832500Log in for availability1 * 1 ST - 86,50 €
Product Information
- Catalog #AvailabilityPrice
- Replacement stirrer assembly, for IntelliCAL™ LBOD101Replacement stirrer assembly, for IntelliCAL™ LBOD101HACH5850800Log in for availability1 * 1 ST - 272,00 €
Product Information
- Catalog #AvailabilityPrice
- Sensor cap replacement kit incl. sensor cap and iButton, for IntelliCAL LBOD101Sensor cap replacement kit incl. sensor cap and iButton, for IntelliCAL LBOD101HACH5838000Log in for availability1 * 1 ST - 324,00 €
Product Information
- Catalog #AvailabilityPrice
- Replacement shroud kit for rugged probes, incl. protective bell and locking ringReplacement shroud kit for rugged probes, incl. protective bell and locking ringHACH5825900Log in for availability1 * 1 ST - 41,40 €
Product Information
- Catalog #AvailabilityPrice
- Sensor cap replacement kit incl. sensor cap and iButton, for IntelliCAL LDO0101Sensor cap replacement kit incl. sensor cap and iButton, for IntelliCAL LDO0101665-0419Log in for availability1 * 1 ST - 253,00 €