Orders LinkContactUsLinkComponent
Micro-tissue grinders
Micro-tissue grinders
Marketsource
Supplier:   Roth Carl
Micro-tissue grinders
Supplier:  Roth Carl
Marketsource
Certificates
Micro-tissue grinders
3 Options Available Below
Product Details & Documents
These micro-tissue grinders with pestles made of borosilicate glass are for pulverising and homogenising minute quantities.
Product Family Options
Product Information
  • Capacity
    Int.Ø×H
    Availability
    Price
  • ROTH0296.1
    Type G 1 with pestle
    Type G 1 with pestle
    0,01 - 0,025 ml
    ~4×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Marketsource
    Marketsource
  • ROTH0297.1
    Type G 2 with pestle
    Type G 2 with pestle
    0,025 - 0,1 ml
    ~6×40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Marketsource
    Marketsource
  • ROTH0298.1
    Type G 3 with pestle
    Type G 3 with pestle
    0,1 - 1 ml
    ~8×50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Marketsource
    Marketsource