Dialysis membranes, Spectra/Por® 6
Supplier: Roth Carl
Certificates
About this item
Dialysis tubing is made from cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
- Stored at 4 °C
- Temperature stable up to 60 °C
Packaging: Packaged wet in 0,05% sodium azide solution; on rolls.
Frequently Bought Together