Orders LinkContactUsLinkComponent
Covered sponge mops, PadMop™
 :   MICRONOVA
Covered sponge mops, PadMop™
 :  MICRONOVA
Covered sponge mops, PadMop™
Covered sponge mops, PadMop™
This fully padded, brick-style flat mop is designed to apply generous amounts of disinfectant to cope with railings, shelving, door jambs, corners, and large surface areas in pharmaceutical production areas.
  • Dual padded cleaning surfaces for railings, door jambs, and corners
  • 19,05 mm urethane foam interiors
  • Holds up to common disinfectants and cleaning agents
  • Launderable and reusable
  • Autoclavable
Fully encased in a choice of low-particle fabric:
NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal.
PolySorb - 100% textured polyester for removing residues and dispersing disinfectant.
The Micronova PadMop™ Covered Sponge Mop features dual cleaning surfaces for generous dispersement of solution on primary surfaces as well as using the top of the pad for cleaning door jambs, railings, piping, and more. The PadMop™ fits onto Micronova’s 360° swivel SlimLine™ adapter, or the LyoTool™ for cleaning lyophiliser shelving.
 : All components must be ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova.
Product Family Options
PadMop™
  • Description
    Dimensions
  • PadMop™ covered sponge mop, NovaPoly™, 100% smooth knit polyester, urethane foam interior
    12.7×35 cm (5×14")
    1 * 1 ST
    1 * 1 ST - 34,00 €
    34,00 €
    1 * 1 ST
       34,00 €
  • PadMop™ covered sponge mop, NovaPoly™, irradiated, 100% smooth knit polyester, urethane foam interior
    12.7×35 cm (5×14")
    1 * 12 ST
    1 * 12 ST - 506,00 €
    506,00 €
    1 * 12 ST
       506,00 €
SlimLine™ Mop Adapters
  •  
    Description
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    1 * 1 ST
    1 * 1 ST - 379,00 €
    379,00 €
    1 * 1 ST
       379,00 €
  •  
    Description
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
    1 * 1 ST
    1 * 1 ST - 395,00 €
    395,00 €
    1 * 1 ST
       395,00 €
Stainless Steel Handles
  •  
    Description
    Length
  • Stainless steel quick disconnect extendable handle
    1575 − 3048 mm
    1 * 1 ST
    1 * 1 ST - 389,00 €
    389,00 €
    1 * 1 ST
       389,00 €
  •  
    Description
    Length
  • Stainless steel quick disconnect handle
    1295 cm
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  •  
    Description
    Length
  • Stainless steel quick disconnect extendable handle
    965 - 1829 mm
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
  •  
    Description
    Length
  • Stainless steel quick disconnect handle
    155 cm
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  •  
    Description
    Length
  • Stainless steel quick disconnect handle
    203 mm
    1 * 1 ST
    1 * 1 ST - 98,70 €
    98,70 €
    1 * 1 ST
       98,70 €
Plastic Handles
  •  
    Description
    Length
  • Plastic ACME threaded handle, with white hand grip
    1219 mm
    1 * 1 ST
    1 * 1 ST - 42,70 €
    42,70 €
    1 * 1 ST
       42,70 €
  •  
    Description
    Length
  • Plastic ACME threaded handle, with white hand grip
    152 cm
    1 * 1 ST
    1 * 1 ST - 46,90 €
    46,90 €
    1 * 1 ST
       46,90 €
  •  
    Description
    Length
  • Plastic ACME threaded extendable handle, with white hand grip
    102 - 183 cm
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  •  
    Description
    Length
  • Plastic ACME threaded extendable handle, with white hand grip
    1016 - 2565 mm
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €