Orders LinkContactUsLinkComponent
Brushes, FlexBrush™
Brushes, FlexBrush™
   Sustainable Options Available
Supplier:   MICRONOVA
Brushes, FlexBrush™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Brushes, FlexBrush™
6 Options Available Below
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications for Product Family
Specifications
  • Catalog No:
  • 129-4202
  • MICRBRWM-22512S_U
  • 129-1068
  • 141-1016
  • 141-1644
  • 141-1645
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
Brushes
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • 129-4202
    J-Brush adapter, hook-shaped brush, stainless steel with plastic handle
    J-Brush adapter, hook-shaped brush, stainless steel with plastic handle
    Nylon
    57,1 mm
    Plastic
    406 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,20 €
    68,20 €
    1 * 1 ST
       68,20 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-4202
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRBRWM-22512S_U
    S-Brush adapter, S-shaped brush, stainless steel with plastic handle
    S-Brush adapter, S-shaped brush, stainless steel with plastic handle
    Nylon
    57,1 mm
    Stainless steel
    533 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    MICRBRWM-22512S_U
    Environmentally Preferable
  • 129-1068
    Adapter, hook-shaped brush, twisted zinc with plastic handle
    Adapter, hook-shaped brush, twisted zinc with plastic handle
    Polyester
    Plastic
    292 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,70 €
    47,70 €
    1 * 1 ST
       47,70 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-1068
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Covers
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • 141-1016
    Polyester cover, NovaPoly
    Polyester cover, NovaPoly
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,30 €
    20,30 €
    1 * 1 ST
       20,30 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1016
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1644
    Polyester cover, irradiated, NovaPoly
    Polyester cover, irradiated, NovaPoly
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 328,00 €
    328,00 €
    1 * 12 ST
       328,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1644
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1645
    Microfibre cover (80% Polyester, 20% Polyamide)
    Microfibre cover (80% Polyester, 20% Polyamide)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,90 €
    22,90 €
    1 * 1 ST
       22,90 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1645
    Environmentally Preferable
    Waste Reducing
      Waste Reducing