ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard 10 mg/L, 12 Elements in Nitric Acid 2% / Hydrofluoric Acid < 0.1% for ICP-MS
VWR® ARISTAR®, Multi-Element Quality Control Standard 10 mg/L, 12 Elements in Nitric Acid 2% / Hydrofluoric Acid < 0.1% for ICP-MS
  84794.180
 :  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard 10 mg/L, 12 Elements in Nitric Acid 2% / Hydrofluoric Acid < 0.1% for ICP-MS
  84794.180
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 10 mg/L {Hf; Ge; Mo; Nb; Sb; Si; Sn; Ta; Te; Ti; W; Zr} in nitric acid 2% / Hydrofluoric acid < 0.1% ARISTAR® for ICP-MS
12 elements at 10 mg/L: Hf, Ge, Mo, Nb, Sb, Si, Sn, Ta, Te, Ti, W, Zr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
 : Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
 : Supplied with certificate of analysis.