Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard, 10 mg/L for 36 Elements in 2% Nitric Acid , for ICP-MS
VWR® ARISTAR®, Multi-Element Quality Control Standard, 10 mg/L for 36 Elements in 2% Nitric Acid , for ICP-MS
Catalog # 84793.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard, 10 mg/L for 36 Elements in 2% Nitric Acid , for ICP-MS
Catalog # 84793.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 10 mg/L {Al; Ag; As; B; Ba; Ca; Cd; Ce; Co; Cr; Cu; Dy; Er; Eu; Fe; Gd; Ho; K; La; Li; Lu; Mg; Mn; Na; Nd; Ni; P; Pb; Rb; Se; Sm; Sr; Tl; Tm; V; Zn} in nitric acid 2% ARISTAR® for ICP-MS

Specifications

About this item

36 Elements: 10 mg/L: Al, Ag, As, B, Ba, Ca, Cd, Ce, Co, Cr, Cu, Dy, Er, Eu, Fe, Gd, Ho, K, La, Li, Lu, Mg, Mn, Na, Nd, Ni, P, Pb, Rb, Se, Sm, Sr, Tl, Tm, V, Zn

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.