ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 33 Elements, for ICP
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 33 Elements, for ICP
Catalog # 84791.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 33 Elements, for ICP
Catalog # 84791.180
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 100 mg/L {Al; Ag; As; B; Ba; Be; Bi; Ca; Cd; Cs; Co; Cr; Cu; Fe; In; K; Li; Mg; Mn; Mo; Na; Ni; Nb; Pb; Rb; Sb; Se; Sr; Ti; Tl; V; U; Zn} in nitric acid 5% ARISTAR® for ICP
Specifications
About this item
33 elements 100 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.