Orders LinkContactUsLinkComponent
Replacement mini union ferrules, SGE
Replacement mini union ferrules, SGE
Replacement mini union ferrules, SGE
Certificates
Replacement mini union ferrules, SGE
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • 558-0188
    Replacement mini union ferrule, 100% graphite, for 0,45 - 0,53 mm ID columns
    Replacement mini union ferrule, 100% graphite, for 0,45 - 0,53 mm ID columns
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,30 €
    59,30 €
    1 * 10 ST
       59,30 €
  • 558-0189
    Replacement mini union ferrule, 100% graphite, for 0,1 - 0,32 mm ID columns
    Replacement mini union ferrule, 100% graphite, for 0,1 - 0,32 mm ID columns
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,30 €
    59,30 €
    1 * 10 ST
       59,30 €
  • 568-0478
    Replacement mini union ferrule, 15% graphite 85% vespel®, no hole
    Replacement mini union ferrule, 15% graphite 85% vespel®, no hole
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 97,90 €
    97,90 €
    1 * 10 ST
       97,90 €
  • 568-0479
    Replacement mini union ferrule, 15% graphite 85% vespel®, 0,3 mm hole
    Replacement mini union ferrule, 15% graphite 85% vespel®, 0,3 mm hole
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 97,90 €
    97,90 €
    1 * 10 ST
       97,90 €