Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Deactivated injector liners

Large volume injector mineral oil analysis liner, 5 mm, 3,4 mm, 54 mm
  VARICP14988
 :   VARIAN
Deactivated injector liners
  VARICP14988
 :  VARIAN
Deactivated injector liners

Large volume injector mineral oil analysis liner, 5 mm, 3,4 mm, 54 mm

 
1 350,00 €
   
1 * 5 ST
 

These deactivated injector liners are optimised for sensitive and reproducible analysis.

  • Easy-opening storage trays
  • High performance standards
  • Silanisation activation prevents sample degradation
 
  • Ext.Ø:
    5 mm
  • Ø int.:
    3,4 mm
  • Description:
    Large volume injector mineral oil analysis liner
  • Length:
    54 mm
  • Seq:
    01