Standard opening screw thread vials
Supplier: THERMO NATIONAL SCIENTIFIC
Jump to:
About this item
Preferred for use with many carousel tray autosamplers. Glass vials are manufactured from superior quality borosilicate clear glass Type 1.
- Re-usable sleeves
Microsampling and high recovery vials allow maximum sample extraction without need for separate inserts.
3 Options Available Below
Product Details & Documents
Preferred for use with many carousel tray autosamplers. Glass vials are manufactured from superior quality borosilicate clear glass Type 1.
- Re-usable sleeves
Microsampling and high recovery vials allow maximum sample extraction without need for separate inserts.
Product Family Options
Screw thread vials
Product Information
- CapacityBaseØ×HMaterialColourCap includedCap SizeAvailabilityPrice
- NSCAC4013-1Vials, without writing patchVials, without writing patch2,00 mlFlat bottom12×32 mmGlassClearNo8-425Log in for availability1 * 100 ST25,30 €
- NSCAC4013-2Vials, without writing patchVials, without writing patch2,00 mlFlat bottom12×32 mmGlassAmberNo8-425Log in for availability1 * 100 ST29,80 €
- NSCAC4013-11MicroVial