Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®
Supplier:   DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®
10 Options Available Below
Product Details & Documents
For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.
Ordering information: Without cap.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Availability
    Price
  • 8,00 ml
    17×60 mm
    Amber
    No
    15-425
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 223,00 €
    223,00 €
    1 * 200 ST
       223,00 €
  • 20,00 ml
    28×57 mm
    Amber
    No
    24-400
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 343,00 €
    343,00 €
    1 * 200 ST
       343,00 €
  • 2,00 ml
    12×35 mm
    Clear
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 156,00 €
    156,00 €
    1 * 200 ST
       156,00 €
  • 4,00 ml
    15×45 mm
    Clear
    No
    13-425
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 164,00 €
    164,00 €
    1 * 200 ST
       164,00 €
  • 8,00 ml
    17×60 mm
    Clear
    No
    15-425
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 193,00 €
    193,00 €
    1 * 200 ST
       193,00 €
  • 12,00 ml
    19×65 mm
    Clear
    No
    15-425
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 224,00 €
    224,00 €
    1 * 200 ST
       224,00 €
  • 16,00 ml
    21×70 mm
    Clear
    No
    18-400
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 232,00 €
    232,00 €
    1 * 200 ST
       232,00 €
  • 24,00 ml
    23×85 mm
    Clear
    No
    20-400
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 316,00 €
    316,00 €
    1 * 200 ST
       316,00 €
  • 20,00 ml
    28×57 mm
    Clear
    No
    24-400
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 383,00 €
    383,00 €
    1 * 200 ST
       383,00 €
  • 40,00 ml
    28×95 mm
    Clear
    No
    24-400
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 437,00 €
    437,00 €
    1 * 200 ST
       437,00 €