Orders LinkContactUsLinkComponent
ATAS optic liners
ATAS optic liners
ATAS optic liners
Certificates
ATAS optic liners
4 Options Available Below
Product Details & Documents
Deactivated liners for ATAS optic injectors.
Product Family Options
Product Information
  • Ext.Ø
    Ø int.
    Length
    Availability
    Price
  • 568-0634
    Split/splitless FocusLiner
    Split/splitless FocusLiner
    5 mm
    3,0 mm
    81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,20 €
    42,20 €
    1 * 1 ST
       42,20 €
  • 568-0633
    Split/splitless FocusLiner
    Split/splitless FocusLiner
    5 mm
    3,0 mm
    81 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 166,00 €
    166,00 €
    1 * 5 ST
       166,00 €
  • 568-0636
    Frit and tapered liner
    Frit and tapered liner
    5 mm
    3,0 mm
    81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 568-0635
    Frit and tapered liner
    Frit and tapered liner
    5 mm
    3,0 mm
    81 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 232,00 €
    232,00 €
    1 * 5 ST
       232,00 €