ContactUsLinkComponent
Air sampler, MAS-100 NT®
Air sampler, MAS-100 NT®
Supplier:  Merck
Certificates
undefined
Air sampler, MAS-100 NT®
Supplier:  Merck
Specifications
  • Catalog No:
  • 1.17274.0001
  • 1.09191.0001
  • Flow rate:
  • 100 L/min
  • 100 L/min
  • Sampling head:
  • Ø 10 cm including 300 holes of Ø 0,6 mm
  • Ø 10 cm including 300 holes of Ø 0,6 mm
  • Material:
  • Aluminium alloy
  • Aluminium alloy
  • Alarm:
  • Up to 100 events
  • Up to 100 events
  • Display:
  • Graphic illuminated and menu-guided navigation with up and down arrows
  • Graphic illuminated and menu-guided navigation with up and down arrows
  • Power:
  • 6 V
  • 6 V