Chiral columns, ReproSil® Chiral-OM-R
Supplier: DR MAISCH
Certificates
About this item
USP L40 column: cellulose tris-3,5-dimethylphenylcarbamate coated to porous silica particles.
Specifications
- Catalog No:
- 554-2616
- 554-2632
- 554-2617
- 554-2615
- 554-2603
- 554-2633
- 554-2635
- 554-2604
- 554-2634
- 554-2602
- USP code:
- L40
- L40
- L40
- L40
- L40
- L40
- L40
- L40
- L40
- L40