Wine refractometers, Master series
Supplier: Atago
About this item
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
5 Options Available Below
Product Details & Documents
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
Specifications for Product Family
Specifications
- Catalog No:
- ATAG2590
- ATAG2591
- ATAG2592
- ATAG2593
- ATAG2594
- Weight:
- 160 g
- 160 g
- 160 g
- 160 g
- 160 g
Specifications
Product Family Options
Product Information
- ModelRangeResolutionAvailabilityPrice
- ATAG2590Wine refractometerWine refractometerMASTER-TA0 − 25 % EtOH ; 0 − 40 °Bx0,2 % EtOH ; 0,2 % °BxLog in for availability1 * 1 ST210,00 €Specifications:
More
Specifications
Cat. No.ATAG2590Weight160 g - ATAG2591Wine refractometerWine refractometerMASTER-OE0 − 130 °Oe ; 0 − 33 °Bx1 °Oe ; 0,2 °BxLog in for availability1 * 1 ST210,00 €Specifications:
More
Specifications
Cat. No.ATAG2591Weight160 g - ATAG2592Wine refractometerWine refractometerMASTER-GOE30 − 170 °Oe ; 0 − 40 °Bx1 °Oe ; 0,2 °BxLog in for availability1 * 1 ST210,00 €Specifications:
More
Specifications
Cat. No.ATAG2592Weight160 g - ATAG2593Wine refractometerWine refractometerMASTER-KMW0 − 27 °KMW ; 0 − 33 °Bx0,2 °KMW ; 0,2 °BxLog in for availability1 * 1 ST210,00 €Specifications:
More
Specifications
Cat. No.ATAG2593Weight160 g - ATAG2594Wine refractometerWine refractometerMASTER-BAUMÉ0 − 21 °Bé ; 0 − 40 °Bx0,2 °Bé ; 0,2 °BxLog in for availability1 * 1 ST210,00 €Specifications:
More
Specifications
Cat. No.ATAG2594Weight160 g