Order Entry
Belgium
ContactUsLinkComponent
VWR®, Standard solution (500 ppm PO₄) for the preparation of phosphate standard solution (5 ppm PO₄) Reag.Ph.Eur. 5002200
VWR®, Standard solution (500 ppm PO₄) for the preparation of phosphate standard solution (5 ppm PO₄) Reag.Ph.Eur. 5002200
Catalog # 88089.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Standard solution (500 ppm PO₄) for the preparation of phosphate standard solution (5 ppm PO₄) Reag.Ph.Eur. 5002200
Catalog # 88089.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    500 mg/l
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Phosphate standard solution Ph Eur N 5002200

Specifications

About this item

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.