Order Entry
Belgium
ContactUsLinkComponent
 
Bismuth subnitrate solution Reag. Ph. Eur. 1011502
Catalog # 87786.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bismuth subnitrate solution Reag. Ph. Eur. 1011502
Catalog # 87786.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :1461.99 g/mol
CAS Number:1304-85-4
Formula:Bi₅O(OH)₉(NO₃)₄
MDL Number:MFCD00064844