Order Entry
Belgium
ContactUsLinkComponent
VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
Catalog # 85739.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
Catalog # 85739.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Ammonium chloride, Reag. Ph.Eur.
  • pH value:
    10.0

Specifications

About this item

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.

  • Ph Eur N 4007300
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.