ContactUsLinkComponent
Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Catalog # 1.16755.1000
Supplier:  Merck
CAS Number:  
Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Catalog # 1.16755.1000
Supplier:  Merck
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel
  • M-Clarity quality level:
    MQ100
Specifications
About this item
10 g/L CsCl + 100 g/L La in dilute hydrochloric acid.