Order Entry
Belgium
ContactUsLinkComponent
Spectromelt® A 1000 Li₂B₄O₇ + 0.07% LiBr for XRF (X-ray fluorescence), Supelco®
Spectromelt® A 1000 Li₂B₄O₇ + 0.07% LiBr for XRF (X-ray fluorescence), Supelco®
Supplier:  Merck
Certificates
Spectromelt® A 1000 Li₂B₄O₇ + 0.07% LiBr for XRF (X-ray fluorescence), Supelco®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (acidimetric):
    ≥ 99.0 %
  • Phosphate (PO₄):
    ≤ 20 ppm
  • Silicate (SiO₂):
    ≤ 150 ppm
  • Sulphate (SO₄):
    ≤ 50 ppm
  • Ag (Silver):
    ≤ 5 ppm
  • Al (Aluminium):
    ≤ 10 ppm
  • Ba (Barium):
    ≤ 5 ppm
  • Ca (Calcium):
    ≤ 20 ppm
  • Cd (Cadmium):
    ≤ 5 ppm
  • Co (Cobalt):
    ≤ 5 ppm
  • Cr (Chromium):
    ≤ 5 ppm
  • Cu (Copper):
    ≤ 5 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • Ga (Gallium):
    ≤ 5 ppm
  • K (Potassium):
    ≤ 10 ppm
  • Mg (Magnesium):
    ≤ 5 ppm
  • Mn (Manganese):
    ≤ 5 ppm
  • Na (Sodium):
    ≤ 20 ppm
  • Ni (Nickel):
    ≤ 5 ppm
  • Pb (Lead):
    ≤ 5 ppm
  • Sr (Strontium):
    ≤ 5 ppm
  • Zn (Zinc):
    ≤ 5 ppm
  • Loss on ignition (1000 °C):
    ≤ 1.0 %
  • Bulk density:
    about 60

Specification Test Results

Related Information

Frequently Bought Together