Order Entry
Belgium
ContactUsLinkComponent
Magnesium nitrate (≥10,0 ± 0,2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Corrosive
Magnesium nitrate (≥10,0 ± 0,2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Catalog # 1.05813.0050
Supplier:  Merck
CAS Number:  
Corrosive
Magnesium nitrate (≥10,0 ± 0,2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Catalog # 1.05813.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Ag (Silver):
    ≤ 0.000002 g/l
  • As (Arsenic):
    ≤ 0.00001 g/l
  • Cd (Cadmium):
    ≤ 0.000001 g/l
  • Pb (Lead):
    ≤ 0.00001 g/l
  • Se (Selenium):
    ≤ 0.0001 g/l
  • Tl (Thallium):
    ≤ 0.00001 g/l

Specification Test Results

About this item

 :
Density:1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8,II
 :M-Clarity™ quality level = MQ100
 :GHS05
CAS Number:10377-60-3
 :13446-18-9
Formula:Mg(NO₃)₂