ContactUsLinkComponent
Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel, Supelco®
Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel, Supelco®
Catalog # 1.02037.1000
Supplier:  Merck
CAS Number:  
Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel, Supelco®
Catalog # 1.02037.1000
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel for the determination of sodium, potassium and lithium by flame spectrometry (enth./cont. 50 g/l CsCl + 250 g/l Al(NO₃)₃ * 9 H₂O)
  • M-Clarity quality level:
    MQ100