Orders LinkContactUsLinkComponent
Infusion filter, Intrapur® Plus
Infusion filter, Intrapur® Plus
Infusion filters, 0,2 µm
Catalog # BRAU4183916
Supplier:   B. Braun
Infusion filter, Intrapur® Plus
Catalog # BRAU4183916
Supplier:  B. Braun
Infusion filter, Intrapur® Plus
Infusion filter, Intrapur® Plus
Infusion filters, 0,2 µm
Price
1 430,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Intrapur plus offers reliable air venting regardless of position with connection lines at both sides of the filter with Luer lock fittings.
  • High throughput
  • PVC free
  • Positively charged membrane
Caution: Should not be used for more than 96 hours.
Specifications
  • Membrane:
    0,2 µm
  • Description:
    Infusion filters