

Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 85.0 %
- ACS - Assay (as H₃PO₄) (by acidimetry):≥85.0 %
- ACS - Antimony (Sb):≤15.000 ppm
- ACS - Arsenic (As):≤1.000 ppm
- ACS - Calcium (Ca):≤0.002 %
- ACS - Chloride (Cl):≤3 ppm
- ACS - Color (APHA):≤10
- ACS - Heavy Metals (as Pb):≤10 ppm
- ACS - Insoluble Matter:≤0.001 %
- ACS - Iron (Fe):≤6 ppm
- ACS - Magnesium (Mg):≤0.002 %
- ACS - Manganese (Mn):≤0.500 ppm
- ACS - Nitrate (NO₃):≤5 ppm
- ACS - Potassium (K):≤6.000 ppm
- ACS - Reducing Substances (mL):≤1.0
- ACS - Sodium (Na):≤10.000 ppm
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Volatile Acids (as CH₃COOH):≤0.001 %
Specification Test Results
Frequently Bought Together