Hydrobromic acid 30% (w/w) ∼5.1 mol/L in acetic acid for peptide research
Supplier: TCI
Certificates
About this item
Synonyms: Hydrogen bromide
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Merck Index:14,04778
Density:1,38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-113-0
UN:2920
ADR:8,II
:GHS02
:GHS05
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Concentration (Argentmetric titration):30.0 to 33.0 w/w%
Specification Test Results
Frequently Bought Together