0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Enzymes
Glucose oxidase (from Aspergillus niger)
Glucose oxidase (from Aspergillus niger)
TCIAG0050-1G
:
TCI
:
Glucose oxidase (from Aspergillus niger)
TCIAG0050-1G
:
TCI
:
Pack type:
Glass bottle for solids
Pk:
1 g
CAS No.:
9001-37-0
Source:
Aspergillus niger
Storage conditions:
Freezer
MDL No.:
MFCD00131182
Enzyme name:
Glucose oxidase