Orders LinkContactUsLinkComponent
VWR® Label Dots on Rolls
VWR® Label Dots on Rolls
VWR® Label Dots on Rolls
VWR® Label Dots on Rolls
These label dot rolls are made from chemically-inert polyester with a unique adhesive formulated to adhere to laboratory glass and plasticware.
  • Withstand temperatures from –196 to +121 °C
  • No need to laminate
  • Use on tubes, racks and boxes
VWR® labels are ideal for all cryostorage and can also be used in most incubators.
Product Family Options
  • Description
    Material
    Colour
    Dimensions
    Packed
  • Label dots for 1,5 ml tubes
    Polyester
    White
    10 mm
    On roll
    1 * 1.000 ST
    1 * 1.000 ST - 72,60 €
    72,60 €
    1 * 1.000 ST
       72,60 €
  • Label dots for 1,5 ml tubes
    Polyester
    White
    10 mm
    On roll
    1 * 5.000 ST
    1 * 5.000 ST - 299,00 €
    299,00 €
    1 * 5.000 ST
       299,00 €
  • Cryo dots, for 1,5 ml tubes
    Polyester
    White
    10 mm
    1 * 3.840 ST
    1 * 3.840 ST - 196,00 €
    196,00 €
    1 * 3.840 ST
       196,00 €
  • Label dots for 1,5 ml tubes
    Polyester
    Multicolour
    10 mm
    On roll
    1 * 5.000 ST
    1 * 5.000 ST - 338,00 €
    338,00 €
    1 * 5.000 ST
       338,00 €
  • Cryo dots, for 2,0 ml tubes, polyestser
    Polyester
    White
    13 mm
    On roll
    1 * 1.000 ST
    1 * 1.000 ST - 110,00 €
    110,00 €
    1 * 1.000 ST
       110,00 €
  • Label dots for 15 and 50 ml tubes
    Polyester
    White
    19 mm
    1 * 2.160 ST
    1 * 2.160 ST - 225,00 €
    225,00 €
    1 * 2.160 ST
       225,00 €