Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-FFAP
Capillary columns, TRACE™ TR-FFAP
Capillary columns, TRACE™ TR-FFAP
Certificates
Capillary columns, TRACE™ TR-FFAP
Capillary columns, TRACE™ TR-FFAP
7 Options Available Below
Product Details & Documents
TRACE™ TR-FFAP GC columns produce outstanding robustness in difficult-to-analyse separations.
  • Engineered specifically for FFAP analysis
  • Polar phase, TPA modified polyethylene glycol
  • Quality tested for acidic compound analysis
  • Equivalent to USP G25 and G35
  • Excellent quality and reproducibility
  • Low bleed even at elevated temperatures
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TR-FFAP
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 462,00 €
    462,00 €
    1 * 1 ST
       462,00 €
  • Capillary
    TR-FFAP
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 766,00 €
    766,00 €
    1 * 1 ST
       766,00 €
  • Capillary
    TR-FFAP
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 817,00 €
    817,00 €
    1 * 1 ST
       817,00 €
  • Capillary
    TR-FFAP
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 300,00 €
    1 300,00 €
    1 * 1 ST
       1 300,00 €
  • Capillary
    TR-FFAP
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 909,00 €
    909,00 €
    1 * 1 ST
       909,00 €
  • Capillary
    TR-FFAP
    0,50 µm
    0,32 mm
    50 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
  • Capillary
    TR-FFAP
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 888,00 €
    888,00 €
    1 * 1 ST
       888,00 €