ContactUsLinkComponent
Staining jar according to Schiefferdecker
  KAVA632499890001
 :  Kavalier
undefined
Staining jar according to Schiefferdecker
  KAVA632499890001
 :  Kavalier
  • Description:
    Staining jars acc. to Schiefferdecker
  • W×D×H:
    90×70×50 mm
  • Pk:
    1
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.