Twisselmann extractors
KAVA632442108090
: Kavalier
- Socket size:29/32 NS
- Description:Complete assembly
- Environmentally Preferable:
- Flask capacity:250 ml
- Pk:1
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Complete assembly for 250 ml flask size.