Orders LinkContactUsLinkComponent
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
Supplier:   Kavalier
Flasks, round bottom, with standard ground joints
Supplier:  Kavalier
Certificates
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
4 Options Available Below
Product Details & Documents
Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
Product Family Options
Product Information
  • Capacity
    Socket size
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    29/32 NS
    145 mm
    85 mm
    Discontinued
  • 250 ml
    29/32 NS
    145 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 567,00 €
    567,00 €
    1 * 10 ST
       567,00 €
  • 500 ml
    29/32 NS
    175 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,30 €
    57,30 €
    1 * 1 ST
       57,30 €
  • 1000 ml
    29/32 NS
    210 mm
    131 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 130,00 €
    130,00 €
    1 * 1 ST
       130,00 €
  • 6000 ml
    60/46 NS
    355 mm
    236 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 355,00 €
    355,00 €
    1 * 2 ST
       355,00 €