Draining racks
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
About this item
Made of wire, plastic coated.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.
2 Options Available Below
Product Details & Documents
Made of wire, plastic coated.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- HECH49027010Draining racks, for 60 test tubes and 5 flasksDraining racks, for 60 test tubes and 5 flasks420×170×610 mmLog in for availability1 * 1 ST128,00 €
- HECH49028010Draining racks, for 12 test tubes and 11 flasksDraining racks, for 12 test tubes and 11 flasks420×170×300 mmLog in for availability1 * 1 ST122,00 €