Orders LinkContactUsLinkComponent
Reversed phase HPLC columns, PRP-C18
Reversed phase HPLC columns, PRP-C18
Supplier:   HAMILTON BONADUZ
Reversed phase HPLC columns, PRP-C18
Supplier:  HAMILTON BONADUZ
Certificates
Reversed phase HPLC columns, PRP-C18
21 Options Available Below
Product Details & Documents
PRP-C18 polymeric HPLC columns are designed to provide high efficiency, reversed phase separations over an extended column life in nearly any mobile phase or pH.
  • Excellent mechanical and thermal stability (>100 °C)
  • LC/MS applications, synthesised DNA, and small molecules
  • Highly cross-linked to prevent shrinkage or swelling
The rigid stationary phase is completely inert to most conditions commonly encountered in reversed phase chromatography. Because of the unique polymer bonding of the C18 to the PS-DVB support, PRP-C18 columns maintain their performance characteristics longer than many traditional C8 or C18 silica columns. These columns are well suited for specialised applications in the clinical, pharmaceutical, environmental, food, forensics and life sciences industries.
Specifications for Product Family
Specifications
  • Catalog No:
  • 554-2538
  • 554-2545
  • 554-2539
  • 554-2546
  • 554-2540
  • 554-2547
  • 554-2541
  • 554-2548
  • 554-2542
  • 554-2549
  • 554-2543
  • 554-2550
  • 554-2544
  • 554-2551
  • 554-2553
  • 554-2552
  • 554-2554
  • HAMI79291
  • 554-2555
  • 554-2556
  • HAMI79918
  • Max. pressure:
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • pH range:
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • Temperature limit:
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • 5 - 85 °C
  • USP code:
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
Specifications
Product Family Options
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    2,10 mm
    50 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 567,00 €
    567,00 €
    1 * 1 ST
       567,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2538
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    2,10 mm
    50 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 608,00 €
    608,00 €
    1 * 1 ST
       608,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2545
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    2,10 mm
    150 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 763,00 €
    763,00 €
    1 * 1 ST
       763,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2539
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    2,10 mm
    150 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 764,00 €
    764,00 €
    1 * 1 ST
       764,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2546
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    2,10 mm
    250 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 824,00 €
    824,00 €
    1 * 1 ST
       824,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2540
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    2,10 mm
    250 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 849,00 €
    849,00 €
    1 * 1 ST
       849,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2547
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    4,60 mm
    50 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2541
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    4,60 mm
    50 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 728,00 €
    728,00 €
    1 * 1 ST
       728,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2548
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    4,60 mm
    150 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 752,00 €
    752,00 €
    1 * 1 ST
       752,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2542
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    4,60 mm
    150 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 862,00 €
    862,00 €
    1 * 1 ST
       862,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2549
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    Stainless steel
    100 Å
    5 µm
    4,60 mm
    250 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 961,00 €
    961,00 €
    1 * 1 ST
       961,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2543
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Analytical
    PRP-C18
    PEEK
    100 Å
    5 µm
    4,60 mm
    250 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 978,00 €
    978,00 €
    1 * 1 ST
       978,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2550
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
  • Preparative
    PRP-C18
    Stainless steel
    100 Å
    12 - 20 µm
    21,20 mm
    250 mm
    PS-DVB functionalised with C18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 300,00 €
    2 300,00 €
    1 * 1 ST
       2 300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2544
    Max. pressure
    5000 psi
    pH range
    1 - 13
    Temperature limit
    5 - 85 °C
    USP code
    L21
Analytical guard columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • 554-2551
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    2,10 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 296,00 €
    296,00 €
    1 * 1 ST
       296,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2551
    Max. pressure
    pH range
    Temperature limit
    USP code
  • 554-2553
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Guard column
    PRP-C18
    PEEK
    12 - 20 µm
    3,00 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 415,00 €
    415,00 €
    1 * 1 ST
       415,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2553
    Max. pressure
    pH range
    Temperature limit
    USP code
  • 554-2552
    Analytical guard column replacement cartridges
    Analytical guard column replacement cartridges
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    2,10 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 400,00 €
    400,00 €
    1 * 5 ST
       400,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2552
    Max. pressure
    pH range
    Temperature limit
    USP code
  • 554-2554
    Analytical guard column replacement cartridges
    Analytical guard column replacement cartridges
    Guard column
    PRP-C18
    PEEK
    5 µm
    3,00 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 367,00 €
    367,00 €
    1 * 5 ST
       367,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2554
    Max. pressure
    pH range
    Temperature limit
    USP code
  • HAMI79291
    Analytical guard column
    Analytical guard column
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    2,10 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 500,00 €
    500,00 €
    1 * 1 ST
       500,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HAMI79291
    Max. pressure
    pH range
    Temperature limit
    USP code
Semi-preparative/preparative guard columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • 554-2555
    Semi-preparative/preparative guard column starter kit (1 holder, 1 cartridge)
    Semi-preparative/preparative guard column starter kit (1 holder, 1 cartridge)
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    4,60 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 394,00 €
    394,00 €
    1 * 1 ST
       394,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2555
    Max. pressure
    pH range
    Temperature limit
    USP code
  • 554-2556
    Semi-preparative/preparative guard column replacement cartridges
    Semi-preparative/preparative guard column replacement cartridges
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    4,60 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 316,00 €
    316,00 €
    1 * 2 ST
       316,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-2556
    Max. pressure
    pH range
    Temperature limit
    USP code
  • HAMI79918
    Semi-preparative/preparative guard column
    Semi-preparative/preparative guard column
    Guard column
    PRP-C18
    Stainless steel
    12 - 20 µm
    4,60 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 500,00 €
    500,00 €
    1 * 1 ST
       500,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HAMI79918
    Max. pressure
    pH range
    Temperature limit
    USP code