Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
Formic acid ~98% (by titrimetric analysis), LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
 
undefined
 
 
 
Formic acid ~98% (by titrimetric analysis), LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
 

 

  • Assay:
    Min. 97.5 %
  • Suitability for the LC-MS:
    complying
  • Non-volatile matter:
    Max. 20 ppm
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Barium (Ba):
    Max. 0.05 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.2 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Manganese (Mn):
    Max. 0.05 ppm
  • Molybdenum (Mo):
    Max. 0.02 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.05 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.02 ppm
  • Thallium (Tl):
    Max. 0.05 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 5 ppm
  • Sulfite (SO₃):
    Max. 10 ppm
  • Ammonium (NH₄):
    Max. 10 ppm
  • APHA:
    Max. 15
  • Appearance of the solution:
    complying
  • Autoignition Temperature:
    1004 °F

 

 

Frequently Bought Together