Order Entry
Belgium
ContactUsLinkComponent
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Irritant
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI
Certificates
undefined
Irritant
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Lambda max.:
    253.0 to 257.0 nm (CH3CN)
  • Absorbance:
    min. 0.280 (0.02g/L, CH3CN, 253.0 to 257.0 nm)

Specification Test Results

Frequently Bought Together