
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Supplier: Novabiochem (Merck biosciences)
Certificates
About this item
CAS Number:56602-33-6
:
Formula:C₁₂H₂₂F₆N₆OP₂
MDL Number:MFCD00011948
:442.28 g/mol
EINECS:260-279-1
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together