Order Entry
Belgium
Orders LinkContactUsLinkComponent
EPPS, 0,2 M buffer solutions

EPPS, 0,2 M buffer solutions

EPPS, 0,2 M buffer solutions
Certificates
EPPS, 0,2 M buffer solutions

4 Options Available Below

Product Details & Documents

CAS No.: 16052-06-5
Physical form: liquid
MDL No.: MFCD00006160

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J61296.AE
  • J61296.AK
  • J61476.AE
  • J61476.AK
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • CAS No.:
  • 16052-06-5
  • 16052-06-5
  • 16052-06-5
  • 16052-06-5
  • MDL No.:
  • MFCD00006160
  • MFCD00006160
  • MFCD00006160
  • MFCD00006160

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pack type
    Pk
    Availability
    Price
  • EPPS buffer solution
    0,2 mol/L
    8,0
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 103,00 €
    103,00 €
    1 * 100 mL
       103,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61296.AE
    Physical form
    Liquid
    CAS No.
    16052-06-5
    MDL No.
    MFCD00006160
  • EPPS buffer solution
    0,2 mol/L
    8,0
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 148,00 €
    148,00 €
    1 * 250 mL
       148,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61296.AK
    Physical form
    Liquid
    CAS No.
    16052-06-5
    MDL No.
    MFCD00006160
  • EPPS buffer solution
    0,2 mol/L
    8,5
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 93,90 €
    93,90 €
    1 * 100 mL
       93,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61476.AE
    Physical form
    Liquid
    CAS No.
    16052-06-5
    MDL No.
    MFCD00006160
  • EPPS buffer solution
    0,2 mol/L
    8,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 136,00 €
    136,00 €
    1 * 250 mL
       136,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61476.AK
    Physical form
    Liquid
    CAS No.
    16052-06-5
    MDL No.
    MFCD00006160