D(+)-Arabitol ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 488-82-4 Molecular Formula: C5H12O5 Formula Weight: 152.15 Melting Point: 100-104° Beilstein Reference: 1720520 Merck Reference: 14,762 Optical Rotation: +132° (c=0.4 in acidified molybdate solution) MDL No.: MFCD00004709
Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Number:488-82-4
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00004709
- Molecular Weight:152.15 g/mol
- Merck Index:14,762
- Melting Pt:100…104 °C
- Storage Temperature:Ambient
- EINECS:207-686-2
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 488-82-4 Molecular Formula: C5H12O5 Formula Weight: 152.15 Melting Point: 100-104° Beilstein Reference: 1720520 Merck Reference: 14,762 Optical Rotation: +132° (c=0.4 in acidified molybdate solution) MDL No.: MFCD00004709
Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Number:488-82-4
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00004709
- Molecular Weight:152.15 g/mol
- Merck Index:14,762
- Melting Pt:100…104 °C
- Storage Temperature:Ambient
- EINECS:207-686-2
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



